Learn More
Attune™ Focusing Fluid (1X)
Attune™ Focusing Fluid (1X)
£29.81 - £160.00
Specifications
Product Type Focusing Fluid
For Use With (Equipment) Attune™ Acoustic Focusing Cytometer
Product Line Attune™
View More Specs
Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10019478
View Documents
Invitrogen
4449791
6 x 1 L
£160.00
Pack of 6
15400984
View Documents
Invitrogen
A24904
10 L
£95.00
10 L
Estimated Shipment: 27-02-2025
to see stock.
Edge
Add to basket
15392496
View Documents
Invitrogen
4488621
1 L
£29.81
1 L
17384843
View Documents Promotion Details
Invitrogen
J106627
20 L
£149.00
20 L
Description
Description
Attune™ Focusing Fluid is an optimized buffer solution that is used in the Attune™ Acoustic Focusing Cytometer for focusing and transportation of particles throughout the fluidics system.
Maintains System Cleanliness
Attune™ Focusing Fluid is a sterile, buffered, azide-free support reagent that contains a surfactant that helps clean the fluidic lines of the Attune™ Acoustic Focusing Cytometer. The Attune™ Focusing Fluid was designed to minimize bubble formation and to prevent build-up of cellular material in the flow cell, resulting in less instrument clogging.
Convenient
The ready-to-use 1X focusing fluid was designed to eliminate the need for dilution prior to use, as well as access to a purified water system. The convenient 10 L packaging provides enough focusing fluid to operate for at least 10 days (the average focusing fluid consumption per day is less than 1 L) and has a spigot to to make filling of the onboard fluidics tanks easier.
High Quality
Attune™ Focusing Fluid is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.
Specifications
Specifications
Focusing Fluid
Attune™ Acoustic Focusing Cytometer
Attune™
Videos
SDS
Documents
Documents
Product Certifications
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Product Title
Attune™ Focusing Fluid (1X)