missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Flow Cytometer, blue/red/violet/green

Designed and developed to remove common barriers associated with flow cytometry, the evolutionary capabilities of the Attune NxT Flow Cytometer

Brand:  Invitrogen™ A29001


Additional Details : Weight : 50.00000kg

Product Code. 17142379

  • POA / Each
Estimated Shipment: 17-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.


Description

Description

  • The 2020 Attune NxT Flow Cytometer has expanded functionalities to drive your research forward. Designed and developed to remove common barriers associated with flow cytometry, the evolutionary capabilities of the Attune NxT Flow Cytometer offer adaptable optical configuration options so you can get the most out of your multicolor analysis.
  • New CytKick and CytKick Max autosampler—more efficiency for high-throughput assays
  • New 21 CFR part 11 compliant software for regulated laboratories on Windows 10 operating system
  • Improved workflow for run protocol and instrument settings management
  • The innovative combination of acoustic focusing technology coupled with traditional hydrodynamic focusing results in acquisition speeds up to 10X faster than other flow cytometers. This technology reduces the time needed to obtain results.
  • True 35,000 event/sec (+10% coincidence rates)
  •  Maximum electronic speed of 65,000 events/sec
  •  Analyze small or limited samples faster than ever
  • Efficiently run traditionally long assays
  • The Attune NxT Flow Cytometer is ideal for immunophenotyping and signaling studies, cell cycle analysis, detection of rare events, stem cell analysis, cancer and apoptosis studies, microbiological assays, and more.
  • The instrument’s modular design provides the flexibility to meet future capability needs with a variety of upgrade options. Purchase what you need now and easily upgrade later. The small footprint of the instrument requires minimal bench space, even allowing use in a cell culture hood.
  • Flexibility—1 to 4 lasers, 14 colors, and 16 parameters
  • Customizable band pass filters for expanded fluorochrome choices
  • The Attune NxT Flow Cytometer easily runs most samples including cardiomyocytes. The system is designed with a clog-resistant syringe-driven system to push the sample with a force of up to 75 psi.
  • Flexibility in sample concentrations and sample types
  • Elimination of time-consuming concentration steps with No-Wash, No-Lyse Filter Kit
  • Analysis of dilute samples, including CSF, stem cells, mouse bone marrow, mouse blood, and thin needle aspirates
  • Acquisition of a high volume of samples in minimal time

Specifications

Specifications

• Attune NxT Flow Cytometer
• Minitower Windows 10 64-bit computer with Attune Cytometric Software
• Power cord with adapters
• USB cable
• Attune Shutdown Solution
• Attune Wash Solution
• Attune Focusing Fluid
• Attune NxT Flow Cell Cleaning Solution
• Attune Performance Tracking Beads
• Quick reference card

488 nm, 405 nm, 637 nm, 532 nm
Attune™ Acoustic Focusing Cytometer
Tube
Attune™
Approved for shipment at Room Temperature or on Wet Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers