Learn More
Invitrogen™ Attune™ NxT Blocker Bar Conversion Kit
The Attune NxT Blocker Bar Conversion Kit provides a 1.5 mm blocking bar for use on the Attune NxT Acoustic Focusing Cytometer, which is delivered with a 0.5 mm blocking bar.
Brand:  Invitrogen™ A35966
Additional Details : Weight : 7.31250kg
Product Code. 17286346
  • £2830.00 / Each
Estimated Shipment: 24-07-2024
to see stock.
Description
Description
  • The Attune NxT Blocker Bar Conversion Kit provides a 1.5 mm blocking bar for use on the Attune NxT Acoustic Focusing Cytometer, which is delivered with a 0.5 mm blocking bar
  • The standard 0.5 mm blocking bar accommodates cells or particles from 0.2–10 microns in size
  • The larger blocking bar included in this kit is recommended for analysis of cells that are primarily larger than 10 microns
  • A larger blocking bar blocks more forward scatter light and allows the detection of larger cells
  • An experienced Thermo Fisher Scientific engineer can install the larger blocking bar on any Attune NxT cytometer, simplifying the instrument settings optimization
Specifications
Specifications
Attune NxT Acoustic Focusing Cytometer
Blocker Bar Conversion Kit
Attune™
• 1 blocker bar
• 1 blocker bar holder
Room Temperature