Learn More
Attune™ NxT Fluorescent Protein Filter Kit
The Attune™ NxT Fluorescent Protein Filter Kit enables the configuration of the Attune™ NxT Acoustic Focusing Cytometer for multiplex detection of GFP, YFP, and mCherry fluorescent proteins.
Brand: Invitrogen 100022775
Code : NEW
Additional Details : Weight : 0.30000kg
Description
Optimized for fluorescent protein detection
At times it can be difficult to detect multiple fluorescent proteins and maintain the best signal detection due to emission signal overlap. This filter kit has been optimized for the detection of GFP in the BL1 channel with a 510/10 BP filter off of the blue laser, YFP in the BL2 channel with a 540/30 BP filter off of the blue laser, and mCherry in the YL2 channel with a 615/25 BP filter off of the yellow laser.
Specifications
Attune Acoustic Focusing Cytometer | |
Contains: • 1 Dichroic 525 LP filter • 1 Emission 510/10 BP filter • 1 Emission 540/30 BP filter • 1 Emission 615/25 BP filter Store (at room temperature.) |
|
Protein Filter Kit | |
Attune™ |
For Research Use Only. Not for use in diagnostic procedures.