missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Shutdown Bottle

Product Code. 15320637
Click to view available options
Quantity:
1 Bottle
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15320637

Brand: Invitrogen™ 100022154

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This is a 175 mL replacement bottle for shutdown solution for the Attune™ NxT Acoustic Focusing Cytometer.

This is a 175 mL replacement bottle for shutdown solution for the Attune™ NxT Acoustic Focusing Cytometer.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Content And Storage Store at room temperature.
Quantity 1 Bottle
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Shutdown Bottle

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.