Learn More
Invitrogen™ Attune™ NxT Waste BTL Tubing
This is replacement tubing for the waste bottle of the Attune NxT Acoustic Focusing Cytometer
Brand:  Invitrogen™ 100096710CST
Product Code. 17156088
  • £63.50 / Each
Explore more special offers
Description
Description
  • This is replacement tubing for the waste bottle of the Attune NxT Acoustic Focusing Cytometer.
  • The tubing can be replaced easily without replacing the entire bottle assembly if the tubing connector to the instrument is broken.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FPL
Each
Attune NxT
Tubing
Room Temperature
Flow Cytometry Accessory