Learn More
Invitrogen™ Attune™ NxT XE3 Computer Upgrade Kit
Designed for use with the Attune NxT Acoustic Focusing Cytometer
Brand:  Invitrogen™ A43668
Product Code. 17186358
  • £4635.00 / Each
Explore more special offers
Description
Description
  • The Invitrogen Attune NxT XE3 Computer Upgrade kit is designed for use with the Attune NxT Acoustic Focusing Cytometer.
  • It includes an OptiPlex XE3 Minitower (Windows 10 Professional 64-bit operating system, 4th Gen Intel Core I7-8700 processor, 32 GB (2x16GB) RAM, 2 GB AMD Radeon video card, 2x2TB SATA hard drive with 64 MB cache, 16X DVD+/- RW drive, network interface card), Dell keyboard (US English), Dell mouse, USB 3.0 Card, USB 3.0 cable, LAN cable, and power cord. Monitor not included.
  • One year from installation for parts, labor, and service travel pursuant to the terms of the Thermofisher Scientific standard limited warranty provisions.
  • One year parts and labor (as provided in written limited warranty statement and accompanying terms in the user manual or other product documentation).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Attune™
1
Computer Upgrade Kit
• OptiPlex XE3 Minitower (Windows 10 Professional 64-bit operating system, 4th Gen Intel Core I7-8700 processor, 32 GB (2x16GB) RAM, 2 GB AMD Radeon video card, 2x2TB SATA hard drive with 64 MB cache, 16X DVD+/- RW drive, network interface card)
• Dell keyboard (US English)
• Dell mouse
• USB 3.0 Card
• USB 3.0 cable
• LAN cable
• Power cord
Monitor not included.
Attune NxT Acoustic Focusing Cytometer