Learn More
Attune™ Shutdown Solution (1X)
Brand: Invitrogen A24975
36.65 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Attune™ Shutdown Solution is a cleaning buffer that is used in the automated shutdown procedure for the Attune™ Acoustic Focusing Cytometer.Minimizes Bubble Formation
This azide-free shutdown solution has been formulated to minimize bubble formulation that can occur in the system lines overnight, to help keep the fluid lines clean, and to minimize microbial growth in the fluid lines.
Use for Instrument Maintenance
The Shutdown cycle is used at the end of each day to ensure that all sample fluids and dyes have been removed from the system, and that the fluidic lines and pumps are filled with Attune™ Shutdown Solution to prevent salt deposits from forming in the lines. Shutdown takes approximately 20 minutes, but most of the steps are automatic and under the control of the Attune™ Cytometric Software. Shutdown procedures should be run at least once a day, even if the instrument is intended for continuous use. Proper cleaning of the instrument ensures its consistent and accurate operation.
Convenient
The ready-to-use 1X shutdown solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system.
High Quality
The Attune™ Shutdown Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality. Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer.

Specifications
Attune™ | |
250 mL | |
Shutdown Solution |
Store at room temperature. | |
Attune Acoustic Focusing Cytometer |
Safety and Handling
- ATTUNE 1X SHUTDOWN SOLUTION
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 3(2H)-isothiazolone,5-chloro-2-methyl-with 2-methyl-3(2H)-isothiazolone
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.