Learn More
Attune™ Wash Solution
Brand:  Invitrogen J24974
 View more versions of this product
Product Code. 17403822
  • £377.00 / 1 L
Explore more special offers
Description
Description
  • Attune™ Wash Solution is an optimized cleaning buffer that is used in the Attune™ Acoustic Focusing Cytometer
  • Helps maintain cleanliness of the fluidics lines to minimize contamination or carryover
  • Formulated to remove cellular debris, prevent protein building up, and minimize dyes sticking to the fluidics lines of the system
  • It is recommended for use during the routine maintenance and automated shutdown procedures of the Attune™ Acoustic Focusing Cytometer
  • The ready-to-use 1X wash solution was designed to eliminate the need for dilution prior to use, as well as access to a purified water system
  • Solution is stable for 30 days after the bottle is opened
  • The Attune™ Wash Solution is prepared, filtered, and packaged in a sterile environment to ensure the highest quality
  • Each batch is traceable by its lot number designation and has been validated for use on the Attune™ Acoustic Focusing Cytometer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at room temperature.
Attune™
Wash Solution
Attune™ Acoustic Focusing Cytometer
1 L