missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Aura™ Particulate Respirator 06900+Gen3 Series
Aura™ Particulate Respirator 06900+Gen3 Series is the next generation of 3M Aura Respirator: FFP protection combined with the original 3-panel, flat-fold design.
£28.70 - £64.24
Specifications
Certifications/Compliance | CE Approved; EN 149:2001+A1:2009 FFP2 NR D |
---|---|
Product Type | Flat Fold |
With Exhalation Valve | Yes |
Quantity | 10/Pk. |
Length (Metric) | 242 mm |
Product Code | Brand | Filter Efficiency | With Exhalation Valve | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Filter Efficiency | With Exhalation Valve | Price | Quantity & Availability | ||||
16270252
|
3M™
7100134422 |
FFP2 | Yes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
16280252
|
3M™
7100134423 |
FFP3 | Yes |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
- New 3M™ Cool Flow™ Comfort Valve allows a greater percentage of warm, exhaled air to escape, helping to increase your comfort
- 3M Advanced Electret (filter) Media helps you breathe easier
- Innovative tabs on the upper and lower panels simplify positioning and adjustment
- Braided headbands are color-coded for quick and easy identification
- Easy to store away with flat-fold design and individual wrapping
- Breathing protection against hazardous work environments
- Made in the UK
Spécification
CE Approved; EN 149:2001+A1:2009 FFP2 NR D | |
Yes | |
242 mm | |
212 mm |
Flat Fold | |
10/Pk. | |
87 mm | |
304 g |