Learn More
3M™ Aura™ Particulate Respirator, Series 9400
Lightweight and foldable. The 3M Aura Particulate Respirator uses a unique three-panel design for greater comfort and easy communication.
£59.60 - £87.69
Products 2
Product Code Brand Filter Type Price Quantity & Availability  
Product Code Brand Filter Type Price Quantity & Availability  
12932505
View Documents
3M™
7000088835
FFP2
£59.60
Pack of 10
In Stock
to see stock.
Edge
Add to basket
12942505
View Documents
3M™
7000088836
FFP3 This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Provide reliable and comfortable protection from a range of airborne disposable hazards
  • Minimize contamination risks to food or pharmaceutical production processes
  • Made without staples or small detachable parts
  • Cool Flow™ exhalation valve option provides effective removal of heat and moisture build up for a more comfortable wear