missing translation for 'onlineSavingsMsg'
Learn More

Heathrow Scientific™ Autoclavable Bags
GREENER_CHOICE

Clear, imprinted Autoclavable Bags with blue biohazard labeling specifically designed for safe disposal.

Brand:  Heathrow Scientific™ HS1002B

 View more versions of this product

Product Code. 16380666

  • £175.00 / Pack of 200
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Constructed of 2.0 mm thick polypropylene.
  • Can be used for repeated autoclaving.
  • Strong side seams securely hold items.
  • Autoclavable at 250°F (121°C).
  • Front write-on panel.
  • 4 inner packs of 50 bags.
  • Disposable.
Specifications

Specifications

Autoclavable Bags
Clear Autoclavable Bags
6.8 gal.
Clear
Hazardous Waste Disposal
Polypropylene
2 mm
50 Pk.
80 mil
16.3 x 23.6 x 0.08 in.
41.5 cm
60 cm
0.02 kg
Room Temperature
Autoclavable (121°C)
25.75 L
Single-use
Autoclave
Non-sterile
200/Cs.
Box
415 x 600 mm
16.3 in.
23.6 in.
0.04 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.