missing translation for 'onlineSavingsMsg'
Learn More

Abeba™ Autoclavable Clogs 39610

Antistatic and autoclavable at 121°C. High slip resistance.

Brand:  Abeba™ 39610/40

63.93 GBP valid until 2024-12-31
Use promo code "21676" to get your promotional price.


 View more versions of this product

Product Code. 15651248

  • £84.55 / 1 pair
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Blue
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications

Specifications

CE EN ISO 20347:2012 OB A, E SRC
40
39610
Blue
Slip-resistant
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers