Learn More
Abeba™ Autoclavable Clogs 9620
Antistatic and autoclavable at 121°C. High slip resistance.
Brand:  Abeba™ 9620/43
 View more versions of this product
Product Code. 15643268
  • £53.65 / 1 pair
Estimated Shipment: 22-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Green
  • Antistatic
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRC
Green
43
9620