missing translation for 'onlineSavingsMsg'
Learn More

Abeba™ Autoclavable Clogs 9630

Antistatic and autoclavable at 121°C. High slip resistance.

Brand:  Abeba™ 9630/39

45.68 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.



Additional Details : Weight : 0.55000kg

 View more versions of this product

Product Code. 15613278

  • £48.08 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket

Description

Description

  • CE EN ISO 20347:2012 OB A, E SRA
  • Autoclavable, sterilisable special material
  • Orange
  • Antistatic
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications

Specifications

CE EN ISO 20347:2012 OB A, E SRA
Orange
39
9630
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers