Learn More
Abeba™ Autoclavable Clogs 9630
Antistatic and autoclavable at 121°C. High slip resistance.
Brand:  Abeba™ 9630/45
Additional Details : Weight : 0.55000kg
 View more versions of this product
Product Code. 15673278
  • £48.08 / 1 pair
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20347:2012 OB A, E SRA
  • Autoclavable, sterilisable special material
  • Orange
  • Antistatic
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRA
Orange
45
9630