Learn More
Cole-Parmer™ Parker Hannifin™ PFA Union Connector Compression Fitting
Compression fittings are an excellent choice to connect rigid tubing systems, able to withstand higher pressures than barbed fittings.
Brand: Cole-Parmer™ 31320-23
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Available in straight, "T", and elbow shapes for unmatched versatility
- Provided with a nut that compresses rigid tubing against the fitting body for high-pressure applications
- Excellent for use with tubing manufactured from fluoropolymers, other rigid plastics and metals
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Sterilization through autoclaving or ethylene oxide (ETO)
Specifications
Compression Fitting | |
PFA | |
1 Each | |
Straight |
Autoclavable | |
3/8 in. tubing O.D. | |
Autoclave or Ethylene Oxide |