Learn More
Cole-Parmer™ Parker Hannifin™ PFA Union Connector Compression Fitting
Compression fittings are an excellent choice to connect rigid tubing systems, able to withstand higher pressures than barbed fittings.
Brand: Cole-Parmer™ 31320-26
This item is not returnable.
View return policy
Description
- Available in straight, "T", and elbow shapes for unmatched versatility
- Provided with a nut that compresses rigid tubing against the fitting body for high-pressure applications
- Excellent for use with tubing manufactured from fluoropolymers, other rigid plastics and metals
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Sterilization through autoclaving or ethylene oxide (ETO)
Specifications
Autoclavable | |
1/4 in. tubing O.D. | |
Autoclave or Ethylene Oxide | |
Compression Fitting |
PFA | |
1 Each | |
Elbow |
Product Content Correction
Product Title
Cole-Parmer™ Parker Hannifin™ PFA Union Connector Compression Fitting > Elbow; For 1/4 in. tubing O.D.
Spot an opportunity for improvement?Share a Content Correction