Learn More
Eppendorf™ Autoclavable Tip-Tub
For aspirating liquids with multi-channel pipettes
Brand: Eppendorf™ 0030058607
25.09 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Tip-Tub reagent reservoir
- 70mL
- Autoclavable reservoir
- Includes 10 reservoirs and 10 lids

Specifications
Autoclavable | |
Aspirating liquids | |
Autoclavable reservoir |
70 mL | |
Multi-channel pipettes | |
10 Pack |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Dairy and Plant-Based Products Testing Promotion
Quality testing products for all your needs.Promo Code: 24134Expires: 2025-03-31
Your input is important to us. Please complete this form to provide feedback related to the content on this product.