missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Microspec™ Autoclave Bags
Designed for contaminated waste disposal in autoclaves or incinerators.
Brand: Microspec™ I1
This item is not returnable.
View return policy
Description
- Clear bags imprinted with blue biohazard labeling designed for safe disposal
- Constructed of thick polypropylene
- Allows for repeated autoclaving
- Strong side seams for securely holding items
- Front write-on panel
Specifications
Autoclave Bags | |
Autoclavable | |
Single-use | |
Non-sterile | |
Polypropylene | |
EcoPak | |
Large | |
633 mm | |
633 mm | |
44.1 lb. |
Room Temperature | |
Clear with Blue Print | |
Polypropylene | |
50 μm | |
500 | |
1200 | |
2 mil | |
24.92 in. | |
840 mm | |
20.1 kg |