Learn More
DuPont™ Micronova™ Autoclave Indicator Tape
Confirms a completed sterilisation cycle in both gravity discharge and vacuum autoclaves. DuPont™ Micronova™ Autoclave Indicator Tape adheres on contact and stays in place through live steam pressure. After use, the tape removes quickly and completely, leaving minimal residue or staining.
Brand:  DuPont™ AUTOPC-1
Product Code. 15596170
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Confirms a completed sterilisation cycle in both gravity discharge and vacuum autoclaves. DuPont™ Micronova™ Autoclave Indicator Tape adheres on contact and stays in place through live steam pressure. After use, the tape removes quickly and completely, leaving minimal residue or staining.
  • Natural background colour, with printed green indicator lines, changing to black when proper levels of steam pressure and temperature have been achieved.
  • Uniquely printed lines cannot be rubbed off and will not contaminate instruments.
  • Saturated crepe paper backing
  • Synthetic polyisoprene adhesive
  • Lead-free ink indicator
  • Standard sterilisation cycle (with pre-vacuum cycle): 121°C (250°F) for 20 minutes, 132° to 134°C (270 to 274°F) for 4 minutes
  • 25mm x 55m length, wound onto a plastic core.
Specifications
Specifications
Polyisoprene, Synthetic
Tape
Sterile
1 Ea.
2.16 in.
1 in.
Natural, Green, Black
Crepe Paper
Gravity discharge and vacuum autoclaves
55 cm x 25.4 cm
55 mm
25.4 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
DuPont™ Micronova™ Autoclave Indicator Tape >