missing translation for 'onlineSavingsMsg'
Learn More

Steris™ 50m Buff Masking Tape

Non-sterile autoclave indicator tape

Brand:  Steris™ 0161

Product Code. 15681934

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Buff color
  • Width: 24 mm
  • Length: 50 m roll
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers