missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPperCHIP™ Autoimmune Epitope Microarray
Autoimmune Epitope Microarray with 4287 epitope specific peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.
Brand: PEPperCHIP™ SPC.042.001
This item is not returnable.
View return policy
Description
- Microarray Content: 4287 B-cell epitopes of the Immune Epitope Database associated with autoimmune diseases including 286 citrullinated peptides as well as the corresponding arginine variants
- Database Epitope Lengths: 3 to 51 aa
- Peptide Lengths: 3 to 17 aa depending on the epitope length; longer epitopes were translated into overlapping 17 aa peptides
- Number of Peptides: 4287 printed in duplicate and HA, polio and c-Myc control peptides
Specifications
PEGMA/PMMA | |
1 Slide | |
1 mm | |
Transparent | |
Individual barcode for each microarray | |
High-density Peptide Microarray | |
Non-sterile |
1 array copy per microarray | |
Combinatorial peptide synthesis by amino acid laser printing | |
ISO 9001:2015 | |
RUO | |
Microscopic Glass | |
Antibody validation, epitope mapping, serum screening, biomarker discovery | |
75.4 x 25 mm |