Learn More
Applied Biosystems™ AutoLys M Tube Locking Lid
This is an extra locking lid for use with the AutoLys M Tube system.
Brand: Applied Biosystems™ A37954
120.00 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Additional Details : Weight : 0.07200kg
Description
Note that two locking lids are included with the purchase of the AutoLys M TubeLifter. The locking lid is required to prevent the inner tube of the AutoLys M Tube from sliding into the outer tube during centrifugation.
Specifications
AutoLys TubeLifter System | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.