Learn More
Applied Biosystems™ AutoLys M Tube Pliers
Centrifugation of the tubes in an AutoLys M Tube Rack is required when processing FFPE samples using the MagMAX FFPE DNA/RNA Ultra Kit.
Brand: Applied Biosystems™ A38261
Additional Details : Weight : 0.12500kg
Descripción
Centrifugation of the tubes in an AutoLys M Tube Rack is required when processing FFPE samples using the MagMAX FFPE DNA/RNA Ultra Kit. The outer-tube will contain the lysate that is processed further to recover the nucleic acids.
Especificaciones
AutoLys TubeLifter System | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.