Learn More
Applied Biosystems™ AutoLys M Tubes and Caps
AutoLys M Tubes are for the preparation of FFPE samples for nucleic acid extraction.
Brand: Applied Biosystems™ A38738
Additional Details : Weight : 1.00000kg
Description
A novel tube-in-tube design creates cleared lysates from FFPE tissue samples, without the need for deparaffinization or wash steps, while increasing DNA and RNA yields. Just add your FFPE sample to the tube, followed by protease buffer and enzyme from the MagMAX FFPE DNA/RNA Ultra Kit. Once digestion is complete, lift the tubes from the AutoLys M Tube Rack using the AutoLys M TubeLifter or AutoLys M Tube Pliers and spin down the samples using a benchtop centrifuge with plate holder (SBS standard format). The FFPE-associated wax and debris are held in the upper chamber of the tube while the lysate passes through. Nucleic acids can then be purified from the clarified lysate using the MagMAX FFPE DNA/RNA Ultra Kit.
Features of the AutoLys M Tube system include:
• FFPE sample lysis and nucleic acid separation from debris in a single tube
• 2-D barcode on bottom tube
• Elimination of use of organic solvents (xylene, CitriSolv) for deparaffinization and washes
• Reduced processing time
• Increased nucleic acid yield
The AutoLys M Tube system is a unique way to process your FFPE samples that will result in improved nucleic acid yields when used in conjunction with the MagMAX FFPE DNA/RNA Ultra Kit, helping ensure the success of downstream assays and reducing the time to data analysis.
Specifications
FFPE Tissue | |
• 25 AutoLys M tubes • 25 AutoLys M caps |
|
AutoLys TubeLifter System | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.