Learn More
Applied Biosystems™ Automated Thermal Cycler (ATC) + Plastics Package, 384-well, EnduraPlates
Applied Biosystems™ Automated Thermal Cycler (ATC) + Plastics Package with EnduraPlates provides reliability and performance while enabling hand-free results.
Brand:  Applied Biosystems™ A41200
7839.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Product Code. 17123189
  • £12060.00 / Each
Explore more special offers
Description
Description
  • Flexible modular design helps save space on a robotic deck
  • The separate control panel module provides flexibility when integrating on a robotic deck
  • The automated lid is compatible with programming for easy, hands-free operation with a liquid handler or plate stacker
  • The heated cover slides forward to cover the plate nest, to help ensure thermal contact and eliminate evaporation
  • The ATC is compatible with full-skirt or half-skirt plates
  • Optimized for performance with Applied Biosystems instruments
  • Certified DNA-, RNAse-, and PCR inhibitor-free with in-process sampling tests
  • Constructed to ANSI/SBS standard, well suited to high-throughput laboratories and robot handling
  • Available in multiple colors for easy organization of laboratory assays
  • Excellent mechanical stability and flatness for your complete workflow needs
TRUSTED_SUSTAINABILITY
Specifications
Specifications
±0.25°C (35°C to 99.9°C)
High-throughput Compatible
Standard
384-well, 20 μL full-skirted plate
No Display
Limited Warranty
USB, Wi-Fi, Ethernet
1 package
384-well Isothermal Block
• Automated Thermal Cycler (ATC), 384-well, 1-meter cable
• 5 x MicroAmp EnduraPlate Optical 384-well Clear Reaction Plates with Barcode (100 plates total)
• 1 x MicroAmp Clear Adhesive Films (100 films)
<0.5°C (20 sec. after reaching 95°C)
12.5 x 7.1 x 5.3 in. (31.7 x 17.9 x 13.3 cm)
100/240 V
Thermal Cycler
4°C to 105°C
2.8°C/sec. (Block), 1.6°C/sec. (Sample)
13.2 lb. (6.0 kg)