Learn More
Bellingham and Stanley™ Automatic Digital Polarimeter
With XPC Technology - Xylem's patented internal Peltier temperature control system
Brand: Bellingham and Stanley™ 37-50
Description
The Automatic Digital Polarimeter features a no maintenance yellow LED and interference filter. When combined with a photodiode detector these technologies allow for the reading of samples of up to 3.0 OD at the commonly used sodium (589 nm) wavelength. ADP400 polarimeters feature angular and sugar scales (ISS) and can be programmed with user scales as well as industry standard methods for displaying invert sugar, inversion (A-B) or when applying other factors such as tube length and concentration. This flexibility makes them ideal for use within laboratories where compliance with Pharmacopoeia is required.
- Accuracy
- ±355
- -255 to 225 via Z sugar scale
- Resolution
- 0.01°A
- 0.001°Z
Specifications
ADP450 Polarimeter |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Bellingham and Stanley - 21 CFR Part 11 Compliance
-
Bellingham and Stanley - ADP450 XPC Polarimeter 37-50