Learn More
BRAND™ Dispenser, tilt measure without bottle
Tilt dispenser is best suited for dispensing suspensions.
Brand:  BRAND™ 43070
Additional Details : Weight : 0.07000kg
 View more versions of this product
Product Code. 10205712
  • £78.30 / Each
Estimated Shipment: 21-08-2024
to see stock.
Add to basket
Description
Description
  • Dispenser head
  • Calibrated to deliver (TD, Ex)
  • Inscriptions in green enamel
  • Particularly well-suited for dispensing suspensions
Specifications
Specifications
Borosilicate Glass 3.3
Without bottle
25 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products