Learn More
BRAND™ Dispenser, tilt measure without bottle
Tilt dispenser is best suited for dispensing suspensions.
Brand:  BRAND™ 43066
Additional Details : Weight : 0.07000kg
 View more versions of this product
Product Code. 10787444
  • £75.39 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Calibrated to deliver (TD, Ex)
  • Inscriptions in green enamel
  • The automatic dispenser is particularly well-suited for dispensing suspensions
  • Specifications
    Specifications
    Borosilicate Glass
    Without bottle
    20 mL
    Product Suggestions
    Product Suggestions
    Videos
    SDS
    Documents
    Documents
    Certificates
    View accessory products