Learn More
Faster™ Automatic Regulator with Full Digital Display and Anemometer
Factory fitted digital display with anemometer results and ability to control UV lamp.
Brand: Faster™ FX0000214980
Code : NEW
Additional Details : Weight : 0.10000kg
Description
- When cabinet is equipped with anemometer with automatic regulation, visualization of air speed and flow alarms will come together
- Cabinet is supplied with keyboard
Specifications
Automatic Regulator | |
FlowFAST V12, V15, V18, SmartFAST Cabinet |