missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms with 384-Channel Disposable Tip Head

Increase pipetting performance with Thermo Scientific™ Tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency.

£2620.00 - £3140.92

Specifications

Filtered Non-filtered
Color Clear
Format Rack
Tip Style Automation
For Use With (Equipment) Agilent/Velocity VPrep, Agilent Bravo
View More Specs

Products 4
Product Code Brand Volume (Metric) Filtered Sterility Price Quantity & Availability  
Product Code Brand Volume (Metric) Filtered Sterility Price Quantity & Availability  
15799077
View Documents
Thermo Scientific™
946-262
10 μL Non-filtered Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15728987
View Documents
Thermo Scientific™
947-262
30 μL Non-filtered Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12924127
View Documents
Thermo Scientific™
948-261
50 μL Non-filtered Sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
16446949
View Documents
Thermo Scientific™
948-262
50 μL Non-filtered Non-sterile This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Increase pipetting performance with Thermo Scientific™ tips for Agilent™ Bravo™ and VPrep™ Automated Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. The result is accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. These automation tips are validated on the actual workstations for which they are created, and are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.

Features:
  • Available as sterile.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications

Specifications

Non-filtered
Rack
Agilent/Velocity VPrep, Agilent Bravo
Clear
Automation
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.