Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head
GREENER_CHOICE
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
£5225.00 - £5695.20
Specifications
Volume (Metric) 30 μL
Color Clear
View More Specs
Products 3
Product Code Brand Sterility Filtered Fit for Purpose Price Quantity & Availability  
Product Code Brand Sterility Filtered Fit for Purpose Price Quantity & Availability  
11711273
GREENER_CHOICE
View Documents
Thermo Scientific™
935-021
Sterile Filtered Sterile and PCR Ready
£5695.20
Pack of 19200
Estimated Shipment: 16-09-2025
to see stock.
Add to basket
11791263
GREENER_CHOICE
View Documents
Thermo Scientific™
935-261
Sterile Non-filtered Sterile
£5365.00
Pack of 19200
Estimated Shipment: 16-09-2025
to see stock.
Add to basket
11701273
GREENER_CHOICE
View Documents
Thermo Scientific™
935-262
Non-sterile Non-filtered Certified Quality
£5225.00
Pack of 19200
Estimated Shipment: 09-09-2025
to see stock.
Add to basket
Description
Description
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).
Specifications
Specifications
30 μL
Clear
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP384 Dispense Head