Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads
GREENER_CHOICE
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
£392.00 - £756.00
Specifications
Color Clear
Format Rack
Tip Style Extended Length
View More Specs
Products 6
Product Code Brand Volume (Metric) Filtered Sterility Fit for Purpose Price Quantity & Availability  
Product Code Brand Volume (Metric) Filtered Sterility Fit for Purpose Price Quantity & Availability  
11781263
GREENER_CHOICE
View Documents
Thermo Scientific™
915-021
20 μL Filtered Sterile Sterile and PCR Ready
£756.00
Pack of 4800
Estimated Shipment: 11-08-2025
to see stock.
Add to basket
11771263
GREENER_CHOICE
View Documents
Thermo Scientific™
915-261
20 μL Non-filtered Sterile Sterile
£441.00
Pack of 4800
Estimated Shipment: 16-09-2025
to see stock.
Add to basket
11781223
GREENER_CHOICE
View Documents
Thermo Scientific™
915-262
20 μL Non-filtered Non-sterile Certified Quality
£392.00
Pack of 4800
Estimated Shipment: 09-09-2025
to see stock.
Add to basket
11741253
GREENER_CHOICE
View Documents
Thermo Scientific™
917-021
130 μL Filtered Sterile Sterile and PCR Ready
£756.00
Pack of 4800
Estimated Shipment: 16-09-2025
to see stock.
Add to basket
11751253
GREENER_CHOICE
View Documents
Thermo Scientific™
917-261
200 μL Non-filtered Sterile Sterile
£441.00
Pack of 4800
Estimated Shipment: 11-08-2025
to see stock.
Add to basket
11791223
GREENER_CHOICE
View Documents
Thermo Scientific™
917-262
200 μL Non-filtered Non-sterile Certified Quality
£392.00
Pack of 4800
In Stock
to see stock.
Add to basket
Description
Description
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384-, and 1536-deep well plates
  • Low percentage coefficient of variation (%CV)
Specifications
Specifications
Clear
Rack
Extended Length
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with MP20 and MP200 Dispense Heads