Learn More
Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Brand: Thermo Scientific™ 923-261
290.27 GBP valid until 2025-12-31
Use promo code "24289" to get your promotional price.
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Caliper™ Zymark™ Liquid Handling Workstations, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Caliper/Zymark RapidPlate, Caliper Presto, Allegro, Zephyr and SciClone.
Features:
Specifications
200 μL | |
Sterile | |
Rack | |
Zymark/Caliper Liquid Handling Workstations | |
Sterile |
Non-filtered | |
Clear | |
Automation | |
Caliper Zymark |