Learn More
Thermo Scientific™ Automation Tips for Qiagen™ Workstations
GREENER_CHOICE
Increase pipetting performance with these automation tips, which undergo a rigorous 15-point quality control program to ensure accuracy and consistency.
Brand:  Thermo Scientific™ 951-261
70.77 GBP valid until 2024-12-31
Use promo code "21899" to get your promotional price.
 View more versions of this product
Product Code. 10274661
  • £119.00 / Pack of 960
Estimated Shipment: 02-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Increase pipetting performance with Thermo Scientific™ Automation Tips for Qiagen™ Universal Pipetting Systems, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance. Compatible with: Qiagen Rosys, Colibri and BioRobot 3000/8000/9600/9604.
Features:
  • Available as sterile and filtered
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
300 μL
Sterile
Rack
96 Tips/Tray, 10 Trays/Case
Qiagen
Non-filtered
Clear
Automation
Universal Fit Pipettor, Qiagen
Sterile