missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
GREENER_CHOICE

Codice prodotto. 10306031
Click to view available options
Volume (Metric):
10 μL
20 μL
50 μL
175 μL
200 μL
1000 μL
Filtered:
Filtered
Non-filtered
For Use With (Equipment):
Cavro, MiniPrep
Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Cavro, Tecan Genesis, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Cavro, Tecan Genesis, Tecan Freedom EVO, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Tecan Aquarius, Tecan TeMo, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Tecan Freedom EVO LiHa Head, Tecan Genesis
Tecan Freedom EVO LiHa Head, Tecan Genesis LiHa Head
Tecan Freedom EVO, MiniPrep, Cavro
Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro
Tecan Genesis, Tecan Freedom EVO, MiniPrep, Cavro, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Fit for Purpose:
Certified Quality
PCR Ready
Sterile
Sterile and PCR Ready
Dimensione della confezione:
Pack of 4800
Pack of 960
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 10306031

Marca: Thermo Scientific™ 904261

Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.

Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.

  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY

Specifica

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Black
Autoclavable Autoclavable
Certifications/Compliance ISO 9001 and ISO 13485
Clean Claims Free of RNase, DNase, DNA, ATP, and endotoxin
Format Rack
Tip Style Automation
Packaging Reload Insert, 96 tips/insert, 10 inserts/pack
Quantity 960 Tips
For Use With (Equipment) Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Fit for Purpose Sterile
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato