Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
GREENER_CHOICE
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Brand:  Thermo Scientific™ 903-251G
118.99 GBP valid until 2025-08-22
Use promo code "24148" to get your promotional price.
 View more versions of this product
Product Code. 10561972
  • ÂŁ144.00 / Pack of 960
Focus on Agri-Science Testing
Use promo code 24136 at checkout
View Available Promotions
Only null left
Estimated Shipment: 11-11-2025
Add to basket
Description
Description
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
  • Available as standard, filtered and/or liquid sensing
  • Certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive, providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 ÎĽL
Sterile
Not Autoclavable
Free of RNase, DNase, DNA, ATP, and endotoxin
Conductive
960 Tips
Sterile
Non-filtered
Black
ISO 9001 and ISO 13485
Rack
Reload Insert, 96 tips/insert, 10 inserts/pack
Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms > Automation tips for Tecan™ Volume: 200ÎĽL; Sterility: Sterile; Color: Conductive; Unit Size: 10 blisters of 96 tips (960 tips in total)