Learn More
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms
GREENER_CHOICE
Benefit from Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms. They increase pipetting performance through the superior tip straightness and low %CV of these pipette tips.
Brand:  Thermo Scientific™ 904-262
106.99 GBP valid until 2025-05-09
Use promo code "24143" to get your promotional price.
 View more versions of this product
Product Code. 10729601
  • £131.00 / Pack of 960
Estimated Shipment: 31-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Get accuracy and precision with Thermo Scientific™ Sterile Automation Tips for Tecan™ Liquid Handling Platforms, which undergo a rigorous 15-point quality control program to ensure consistency. Their superior tip straightness and a low coefficient of variation provide accurate and precise performance.
  • Available nonsterile with-or-without Thermo Scientific™ ART™ filter technology
  • Available in filtered, low retention or extended length for increased volume to reach into deep well plates
  • Lot-tested during production on actual workstation for which they are created
  • Fully conductive providing increased pipetting performance
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1000 μL
Non-sterile
Not Autoclavable
Free of RNase, DNase, DNA, ATP, and endotoxin
Low Retention, Extended Length
960 Tips
Certified Quality
Non-filtered
Clear
ISO 9001 and ISO 13485
Rack
Reload Insert, 96 tips/insert, 10 inserts/pack
Cavro, MiniPrep, PerkinElmer MultiProbe II HT, PerkinElmer MultiProbe II HT EX, PerkinElmer Janus
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
Product Content Correction
Product Title
Thermo Scientific™ Automation Tips for Tecan™ Liquid Handling Platforms > Automation tips for Tecan™ Volume: 1000μL; Sterility: Sterile; Color: Clear; Unit Size: 10 blisters of 96 tips (960 tips in total)