Learn More
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
Increase pipetting performance with these automation tips. Each pipette tip undergoes a rigorous 15-point quality control program to ensure consistency and accuracy achieved by superior tip straightness and a low coefficient of variation.
Brand: Thermo Scientific™ 965-021
657.89 GBP valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
Features:
- Available in nonsterile, sterile and sterile filtered
- Validated on actual workstation for which tip is created
- Low percentage coefficient of variation
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Specifications
50 μL | |
Sterile | |
Rack | |
Eppendorf epMotion | |
Sterile and PCR Ready |
Filtered | |
Clear | |
Automation | |
Eppendorf epMotion |