missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System
GREENER_CHOICE

Increase pipetting performance with these automation tips. Each pipette tip undergoes a rigorous 15-point quality control program to ensure consistency and accuracy achieved by superior tip straightness and a low coefficient of variation.

£410.00 - £913.00

Specifications

Color Clear
Format Rack
For Use With (Equipment) Eppendorf epMotion
Product Line Eppendorf epMotion
View More Specs

Products 8
Product Code Brand Volume (Metric) Filtered Sterility Fit for Purpose Price Quantity & Availability  
Product Code Brand Volume (Metric) Filtered Sterility Fit for Purpose Price Quantity & Availability  
13326048
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
965-021
50 μL Filtered Sterile Sterile and PCR Ready
£692.52
Pack of 3840
Estimated Shipment: 01-07-2025
to see stock.
Add to basket
 
15227626
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
965-262
50 μL Non-filtered Non-sterile Certified Quality
£422.00
Pack of 3840
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
15237626
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
966-021
300 μL Filtered Sterile Sterile and PCR Ready
£699.33
Pack of 3840
In Stock
to see stock.
Add to basket
 
15257626
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
966-261
300 μL Non-filtered Sterile Sterile
£492.00
Pack of 3840
Estimated Shipment: 28-07-2025
to see stock.
Add to basket
 
15267626
GREENER_CHOICE
View Documents
Thermo Scientific™
966-262
300 μL Non-filtered Non-sterile Certified Quality
£422.00
Pack of 3840
Estimated Shipment: 07-07-2025
to see stock.
Add to basket
 
13306068
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
967-021
1000 μL Filtered Sterile Sterile and PCR Ready
£913.00
Pack of 3840
In Stock
to see stock.
Add to basket
 
13346068
GREENER_CHOICE
View Documents
Thermo Scientific™
967-261
1000 μL Non-filtered Sterile Sterile
£475.00
Pack of 3840
Estimated Shipment: 11-08-2025
to see stock.
Add to basket
 
13356068
GREENER_CHOICE
View Documents Promotion Details
Thermo Scientific™
967-262
1000 μL Non-filtered Non-sterile Certified Quality
£410.00
Pack of 3840
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
 
Description

Description

Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System are available in non-sterile, sterile and sterile filtered tips. Each pipet tip undergoes a rigorous 15-point quality control program to ensure consistency, thus providing accurate and precise performance achieved by superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created and the sterile pipette tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.

Features:
  • Available as sterile and filtered.
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen.
  • Lot-tested during production on actual workstation for which they are created.
  • Low percentage coefficient of variation (%CV).

Warranty:A five (5)-year shelf life from the date of manufacture is recommended.
Specifications

Specifications

Clear
Rack
Eppendorf epMotion
Eppendorf epMotion
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Automation Tips for the Eppendorf™ epMotion™ Automated Pipetting System

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.