missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Autophagy Inhibitor, 3-MA

Product Code. 15406599
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
This item is not returnable. View return policy

Product Code. 15406599

Brand: Thermo Scientific Chemicals J64813.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5142-23-4 | C6H7N5, C6H7N5 | 149.16 g/mol

A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5142-23-4
Molecular Formula C6H7N5, C6H7N5
Synonym 3-Methyladenine; 6-Amino-3-methylpurine

Specifications

Quantity 25 mg
Description White
Content And Storage 2°C to 8°C
For Use With (Application) A synthetic intermediate and a cell-permeable autophagic sequestration blocker that protects cerebellar granule cells from apoptosis post serum/potassium deprivation
Formula Weight 149.15g/mol
Product Type Signal Transduction Reagent
Form Solid
MDL Number MFCD00010531
Beilstein 146087
InChI Key FSASIHFSFGAIJM-UHFFFAOYSA-N
SMILES CN1C=NC(N)=C2N=CN=C12
IUPAC Name 3-methyl-3H-purin-6-amine
Molecular Weight (g/mol) 149.16
Show More Show Less
compliance-icons
Product Identifier
  • Autophagy Inhibitor, 3-MA
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : 3-Methyl-3H-adenine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.