Learn More
Trajan Scientific and Medical™ Autosampler Syringes with Fixed Needle for Thermo Scientific GCs
Gives better performance and superior quality. Trajan Scientific and Medical Autosampler Syringes with Fixed Needle for Thermo Scientific GCs has the knowledge and expertise to manufacture an SGE syringe.
Brand:  Trajan Scientific and Medical™ 0019811
 View more versions of this product
Product Code. 15248443
  • £89.40 / Each
Explore more special offers
Description
Description
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications
Specifications
CTC/Thermo (Classic Button) Syringe
0.63 mm
5 μL
Cone
23 G
Fixed Needle
50 mm
Thermo Gas Chromatography