missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Avidin, AP conjugate
Avidin, AP conjugate
Brand: Molecular Probes™ 434422
169.48 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Reactivity: AP-Avidin reacts with biotin.Applications: Immunohistochemistry, ELISA
Specifications
AP | |
Store in refrigerator (2–8°C). | |
Liquid |
2 mL | |
Wet Ice | |
AP (Alkaline Phosphatase) |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction