missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Avidin, Egg White
Avidin, egg white, 5 mg
£62.00 - £499.00
Specifications
Conjugate | Unconjugated |
---|---|
Content And Storage | Store in freezer (-5 to -30°C). |
Shipping Condition | Room Temperature |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11554037
|
Molecular Probes™
A2667 |
5 mg |
£62.00
5mg |
Estimated Shipment: 26-07-2024
Log in to see stock. |
|||||
11564137
|
Molecular Probes™
A887 |
100 mg |
£499.00
100mg |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Avidin from egg white is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.Specifications
Unconjugated | |
Store in freezer (-5 to -30°C). | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.