missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Avidin, FITC conjugate

Product Code. p-7075642
Click to view available options
Unit Size:
5mg
Each
Packaging:
1 Each
This item is not returnable. View return policy

Product Code. 10009232

Brand: Invitrogen™ 434411

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Avidin, FITC conjugate

Our avidin, fluorescein conjugate has excitation and emission maxima ∼494/518. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate FITC (Fluorescein)
Content And Storage Store in refrigerator (2–8°C).
Shipping Condition Wet Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.