missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Avidin, NeutrAvidin™, Horseradish Peroxidase conjugate

Product Code. 10053132
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10053132

Brand: Invitrogen™ A2664

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Avidin, NeutrAvidin™, Horseradish Peroxidase conjugate

Our horseradish peroxidase (HRP) conjugate of NeutrAvidin™ biotin-binding protein - a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point - can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate HRP
Quantity 1 mg
Content And Storage Store in freezer (-5 to -30°C).
Shipping Condition Wet Ice
Product Line NeutrAvidin
Label or Dye HRP (Horseradish Peroxidase)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.