missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Avidin, NeutrAvidin™, Oregon Green™ 488 conjugate

Product Code. 10564803
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10564803

Brand: Invitrogen™ A6374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Avidin, NeutrAvidin™, Oregon Green™ 488 conjugate

Our Oregon Green™ 488 conjugate of NeutrAvidin™ biotin-binding protein - a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point - can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding. This bright, green-fluorescent reagent (absorption/emission maxima ∼496/524 nm) is ideal for detecting biotinylated targets in fluorescence microscopy and flow cytometry applications.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Oregon Green 488
Quantity 1 mg
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
Product Line NeutrAvidin, Oregon Green

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.