missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Avidin, NeutrAvidin™, Texas Red™ conjugate
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Our Texas Red™ conjugate of NeutrAvidin™ biotin-binding protein - a form of avidin that has been processed to remove carbohydrate and to lower its isoelectric point - can substantially decrease background due to nonspecific binding. The method used to deglycosylate the avidin retains its specific binding. This bright, red-fluorescent reagent (absorption/emission maxima ∼595/615 is ideal for detecting biotinylated targets in fluorescence microscopy applications.
Specifications
Specifications
| Conjugate | Texas Red |
| Quantity | 1 mg |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Shipping Condition | Room Temperature |
| Product Line | NeutrAvidin, Texas Red |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction