missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AW Standard Super-Cel™ NF Diatomaceous Earth, Honeywell Fluka™
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 250 g |
Packaging | Poly Bottle |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Aw Standard Super-Cel™ Nf |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'